Läs mer
Primidone, TRC
Produktkod. 30409700
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30409700 10 g
1 options
Produktkod. 30409700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP73400010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Primidone
CAS 125-33-7
Molekylformel C12 H14 N2 O2
Kvantitet 10 g
Synonym Primidone, 5-Ethyl-5-phenyldihydropyrimidine-4,6(1H,5H)-dione, 4,6(1H,5H)-Pyrimidinedione','5-ethyldihydro-5-phenyl- (6CI,8CI,9CI), 2-Deoxyphenobarbital, 2-Desoxyphenobarbital, 5-Ethyl-5-phenylhexahydropyrimidine-4,6-dione, 5-Ethyldihydro-5-phenyl-4,6(1H,5H)-pyrimidinedione, 5-Ethylhexahydro-5-phenylpyrimidine-4,6-dione, 5-Phenyl-5-ethyl-hexahydropyrimidine-4,6-dione, Hexamidine, Hexamidine (antiepileptic), Hexamydin, Lepimidin, Liskantin, Milepsin, Misodine, Misolyne, Mizodin, Mizolin, Mylepsin, Mysedon, Mysoline, NSC 41701, Neurosyn, Primaclone, Primacone, Primakton, Primidon, Pyrimidone Medi-pets, Resimatil, Roe 101, Sertan
InChI formel InChI=1S/C12H14N2O2/c1-2-12(9-6-4-3-5-7-9)10(15)13-8-14-11(12)16/h3-7H,2,8H2,1H3,(H,13,15)(H,14,16)
LEDER CCC1(C(=O)NCNC1=O)c2ccccc2
IUPAC-namn 5-ethyl-5-phenyl-1,3-diazinane-4,6-dione
Molekylvikt (g/mol) 218.25
Formel vikt 218.1055