Learn More
Bellingham Stanley™ Print Cable
Product Code. 11900068
Change view
Click to view available options
Voltage:
230 VAC
Unit Size:
Each
Product Code. Voltage Unit Size
11900068 230 VAC Each
1 options
Product Code. 11900068 Supplier Bellingham Stanley™ Supplier No. 5402
  • ONLINE EXCLUSIVE PRICE
    850.00 SEK
    759.31 SEK / Paket med 1
    Save 90.69 SEK
    10% Av
Beräknad leverans: 16-12-2026
Specifications
Type Print Cable
For Use With (Equipment) CBM-910 or BS1012 Serial, RFM900/RFM700 and ADP/ADS Model
Voltage 230 VAC