missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PRKAB1 (Human) Recombinant Protein
Human PRKAB1 full-length ORF ( AAH01007, 1 a.a. - 270 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005564-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGNTSSERAALERHGGHKTPRRDSSGGTKDGDRPKILMDSPEDADLFHSEEIKAPEKEEFLAWQHDLEVNDKAPAQARPTVFRWTGGGKEVYLSGSFNNWSKLPLTRSHNNFVAILDLPEGEHQYKFFVDGQWTHDPSEPIVTSQLGTVNNIIQVKKTDFEVFDALMVDSQKCSDVSELSSSPPGPYHQEPYVCKPEERFRAPPILPPHLLQVILNKDTGISCDPALLPEPNHVMLNHLYALSIKDGVMVLSATHRYKKKYVTTLLYKPI
Specifications
AAH01007 | |
protein kinase, AMP-activated, beta 1 non-catalytic subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRKAB1 | |
GST |
5564 | |
Wheat germ expression system | |
25 μg | |
AMPK, HAMPKb, MGC17785 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |