missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKAR1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17991-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The second messenger cyclic AMP (cAMP) mediates diverse cellular responses to external signals such as proliferation, ion transport, regulation of metabolism and gene transcription by activation of the cAMP-dependent protein kinase (cAPK or PKA). Activation of PKA occurs when cAMP binds to the two regulatory subunits of the tetrameric PKA holoenzyme resulting in release of active catalytic subunits. Three catalytic (C) subunits have been identified that each represent specific gene products. Activation of transcription upon elevation of cAMP levels results from translocation of PKA to the nucleus where it phosphorylates the transcrip- tion factor cAMP response element binding protein (CREB) on serine 133 which in turn leads to TFIIB binding to TATA-box-binding protein TBP1, thus linking phospho-CREB to the pol II transcription initiation complex.Specifications
PRKAR1B | |
Polyclonal | |
Unconjugated | |
PRKAR1B | |
PRKAR1, PRKAR1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19085, 25521, 5575 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12849, P31321, P81377 | |
Prkar1b | |
PRKAR1B Fusion Protein Ag12426 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |