missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11623-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Protein kinase D2 (PKD2) is a novel human serine threonine protein kinase gene. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKC and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.Specifications
PRKD2 | |
Polyclonal | |
Unconjugated | |
PRKD2 | |
HSPC187, PKD2, PRKD2, protein kinase D2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101540, 25865, 292658 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XIS9, Q8BZ03, Q9BZL6 | |
PRKD2 | |
PRKD2 Fusion Protein Ag2192 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |