missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKG1 Recombinant Human Protein
PRKG1 is a cGMP-dependent serine/threonine kinase.
Brand: Invitrogen™ PR8028A
Additional Details : Weight : 0.01000kg
Description
PRKG1 has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation
Specifications
NP_006249 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
PRKG1, PGK | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
105.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PKG Kinase Family | |
Recombinant | |
Baculovirus |