missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKG2 (PKG2) Recombinant Human Protein
PRKG2 (PKG2) exhibits many cellular functions.
Brand: Invitrogen™ PV4277
Additional Details : Weight : 0.00900kg
Description
PRKG2 (PKG2) has cAMP- and cGMP-dependent protein kinase activity.
Spécification
NP_006250 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
PRKG2 (PKG2) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
116.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PKG Kinase Family | |
Recombinant | |
Baculovirus |