Läs mer
PRO-LAD, TRC
Produktkod. 30423701
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30423701 2.5 mg 2.5mg
30415122 25 mg -
30412112 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423701 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8345602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65527-63-1
Molekylformel C22H29N3O
Molekylvikt (g/mol) 351.49
Synonym N-Propylnorlysergic acid N,N-diethylamide, N-PropylnorLSD, (8beta)-9,10-didehydro-N,N-diethyl-6-propylergoline-8-carboxamide
IUPAC-namn (6aS,9S)-N,N-diethyl-7-propyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
LEDER CCCN1C[C@H](C=C2[C@@H]1Cc3c[nH]c4cccc2c34)C(=O)N(CC)CC
Specifikationer
Kemiskt namn eller material PRO-LAD
Kvantitet 2.5 mg
InChI formel InChI=1S/C22H29N3O/c1-4-10-25-14-16(22(26)24(5-2)6-3)11-18-17-8-7-9-19-21(17)15(13-23-19)12-20(18)25/h7-9,11,13,16,20,23H,4-6,10,12,14H2,1-3H3/t16-,20-/m0/s1
Formel vikt 351.2311