Läs mer
Probucol Disuccinate, TRC
Produktkod. 30415166
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30415166 1 g
1 options
Produktkod. 30415166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7551101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Probucol Disuccinate
CAS 216168-45-5
Molekylformel C39H56O8S2
Kvantitet 1 g
Synonym Butanedioic Acid (1-Methylethylidene)bis[thio[2,6-bis(1,1-dimethylethyl)-4,1-phenylene]] Ester, Butanedioic Acid','1,1'-[(1-Methylethylidene)bis[thio[2,6-bis(1,1-dimethylethyl)-4,1-phenylene]]] Ester
InChI formel InChI=1S/C39H56O8S2/c1-35(2,3)25-19-23(20-26(36(4,5)6)33(25)46-31(44)17-15-29(40)41)48-39(13,14)49-24-21-27(37(7,8)9)34(28(22-24)38(10,11)12)47-32(45)18-16-30(42)43/h19-22H,15-18H2,1-14H3,(H,40,41)(H,42,43)
LEDER CC(C)(C)c1cc(SC(C)(C)Sc2cc(c(OC(=O)CCC(=O)O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1OC(=O)CCC(=O)O)C(C)(C)C
IUPAC-namn 4-[2,6-ditert-butyl-4-[2-[3,5-ditert-butyl-4-(3-carboxypropanoyloxy)phenyl]sulfanylpropan-2-ylsulfanyl]phenoxy]-4-oxobutanoic acid
Molekylvikt (g/mol) 716.99
Formel vikt 716.3417