Läs mer
Probucol, TRC
Produktkod. 30426709
Change view
Kvantitet:
100 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426709 5 g
30419565 100 mg
30411643 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7551005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23288-49-5
Molekylformel C31 H48 O2 S2
Molekylvikt (g/mol) 516.84
Synonym Phenol, 4,4'-[(1-methylethylidene)bis(thio)]bis[2,6-bis(1,1-dimethylethyl)-, Acetone, bis(3,5-di-tert-butyl-4-hydroxyphenyl) mercaptole (8CI), Phenol, 4,4'-(isopropylidenedithio)bis[2,6-di-tert-butyl- (8CI), 4,4'-(Isopropylidenedithio)bis[2,6-di-tert-butylphenol], Biphenabid, Bisbid, Bisphenabid, DH 581, Lipomal, Lorelco, Lurselle, NSC 652160, NSC 86225, Panavir, Phenbutol, Probucol, Sinlestal
IUPAC-namn 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
LEDER CC(C)(C)c1cc(SC(C)(C)Sc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
Specifikationer
Kemiskt namn eller material Probucol
Kvantitet 5 g
InChI formel InChI=1S/C31H48O2S2/c1-27(2,3)21-15-19(16-22(25(21)32)28(4,5)6)34-31(13,14)35-20-17-23(29(7,8)9)26(33)24(18-20)30(10,11)12/h15-18,32-33H,1-14H3
Formel vikt 516.3096