Läs mer
Prochloraz Manganese Chloride Complex, TRC
Produktkod. 30138032
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138032 500 mg
30138487 1 g
17974695 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138032 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP755275500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75747-77-2
Molekylformel C60H64Cl14MnN12O8
Molekylvikt (g/mol) 1632.51
Synonym Prochloraz Manganese, Dichlorotetrakis[N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-1H-imidazole-1-carboxamide-κO1]manganese, Prochloraz Manganese(II) Complex (4:1), N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-1H-imidazole-1-carboxamide Manganese Complex
LEDER ClC1=CC(Cl)=C(OCCN(/C(N2C=NC=C2)=O/[Mn+2](/O=C(N3C=CN=C3)/N(CCOC4=C(Cl)C=C(Cl)C=C4Cl)CCC)(/O=C(N5C=CN=C5)/N(CCOC6=C(Cl)C=C(Cl)C=C6Cl)CCC)(/O=C(N(CCOC7=C(Cl)C=C(Cl)C=C7Cl)CCC)/N8C=NC=C8)([Cl-])[Cl-])CCC)C(Cl)=C1
Specifikationer
Kemiskt namn eller material Prochloraz Manganese Chloride Complex
Kvantitet 500 mg
InChI formel InChI=1S/4C15H16Cl3N3O2.2ClH.Mn/c4*1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18;;;/h4*3,5,8-10H,2,4,6-7H2,1H3;2*1H;/q;;;;;;+2/p-2
Formel vikt 1625