Läs mer
Prochloraz, TRC
Produktkod. 30003649
Change view
Kvantitet:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003649 500 mg
30009945 10 g
17962475 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003649 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP755280500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67747-09-5
Molekylformel C15 H16 Cl3 N3 O2
Molekylvikt (g/mol) 376.67
Synonym N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-1H-imidazole-1-carboxamide, BTS 40542, BTS 40542-7877, DMI, KI 835, Mirage, Mirage (pesticide), N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide, Plocloraz, Prelude, Prochloraz, Sportak, Sportake
IUPAC-namn N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide
LEDER CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)n2ccnc2
Specifikationer
Kemiskt namn eller material Prochloraz
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C15H16Cl3N3O2/c1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18/h3,5,8-10H,2,4,6-7H2,1H3
Formel vikt 375.0308
Procent renhet >95%