missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (H20), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543884
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
Specifikationer
| Procollagen I N-Peptide | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02452 | |
| COL1A1 | |
| Recombinant PINP (Gln23-Pro161) expressed in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| H20 | |
| Unconjugated | |
| COL1A1 | |
| P1NP; Procollagen I N | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering