Läs mer
Procyanidin A2, TRC
Produktkod. 30426847
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30426847 100 mg
1 options
Produktkod. 30426847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP755838100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Procyanidin A2
CAS 41743-41-3
Molekylformel C30 H24 O12
Kvantitet 100 mg
Synonym 8,14-Methano-2H,14H-1-benzopyrano[7,8-d][1,3]benzodioxocin-3,5,11,13,15-pentol','2,8-bis(3,4-dihydroxyphenyl)-3,4-dihydro-','(2R,3R,8S,14R,15R)-, 8,14-Methano-2H,14H-1-benzopyrano[7,8-d][1,3]benzodioxocin-3,5,11,13,15-pentol','2,8-bis(3,4-dihydroxyphenyl)-3,4-dihydro-','[2R-(2α,3α,8β,14β,15R*)]-, (+)-Epicatechin-(4β-8,2β-O-7)-epicatechin, (+)-Proanthocyanidin A2, (2R,3R,8S,14R,15R)-2,8-Bis(3,4-dihydroxyphenyl)-3,4-dihydro-8,14-methano-2H,14H-1-benzopyrano[7,8-d][1,3]benzodioxocin-3,5,11,13,15-pentol, Dimeric catechin, Epicatechin-(4β→8,2β→O→7)-epicatechin, Litchiderm IS 9704, Litchiderm LS 9704, Proanthocyandin A-2, Proanthocyanidin A2, Procyanidin A2, Procyanidin dimer A2, Procyanidol A2
InChI formel InChI=1S/C30H24O12/c31-13-7-20(37)24-22(8-13)41-30(12-2-4-16(33)19(36)6-12)29(39)26(24)25-23(42-30)10-17(34)14-9-21(38)27(40-28(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,21,26-27,29,31-39H,9H2/t21-,26-,27-,29-,30+/m1/s1
LEDER O[C@@H]1Cc2c(O)cc3O[C@@]4(Oc5cc(O)cc(O)c5[C@@H]([C@H]4O)c3c2O[C@@H]1c6ccc(O)c(O)c6)c7ccc(O)c(O)c7
Molekylvikt (g/mol) 576.5
Formel vikt 576.1268