Läs mer
Procyanidin B1, TRC
Produktkod. 17912355
Change view
Kvantitet:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912355 1 mg
17992115 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17912355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7558201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20315-25-7
Molekylformel C30 H26 O12
Molekylvikt (g/mol) 578.52
Synonym (2R,2'R,3R,3'S,4R)-2,2'-bis(3,4-dihydroxyphenyl)-3,3',4,4'-tetrahydro-[4,8'-Bi-2H-1-benzopyran]-3,3',5,5',7,7'-hexol, (-)-Epicatechin-(4β-8)-(+)-catechin, Proanthocyanidin B1, Procyanidol B1
IUPAC-namn (2R,3R,4R)-2-(3,4-dihydroxyphenyl)-4-[(2R,3S)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-8-yl]-3,4-dihydro-2H-chromene-3,5,7-triol
LEDER O[C@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4cc(O)cc(O)c34)c5ccc(O)c(O)c5)c2O[C@@H]1c6ccc(O)c(O)c6
Specifikationer
Kemiskt namn eller material Procyanidin B1
Kvantitet 1 mg
InChI formel InChI=1S/C30H26O12/c31-13-7-20(37)24-23(8-13)41-29(12-2-4-16(33)19(36)6-12)27(40)26(24)25-21(38)10-17(34)14-9-22(39)28(42-30(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,22,26-29,31-40H,9H2/t22-,26+,27+,28+,29+/m0/s1
Formel vikt 578.1424