Läs mer
Prodigiosin, MedChemExpress
Produktkod. 16414426
Encompass_Preferred
Change view
Kvantitet:
100 μg
Produktkod. Kvantitet
16414426 100 μg
1 options
Produktkod. 16414426 Leverantör Medchem Express Leverantörsnummer HY100711
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
MedChemExpress Prodigiosin (Prodigiosine) is a red pigment produced by bacteria as a bioactive secondary metabolite. Prodigiosin is a potent inhibitor of the Wnt/β-catenin pathway. Prodigiosin has antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer properties.
Prodigiosin (Prodigiosine) is a red pigment produced by bacteria as a bioactive secondary metabolite. Prodigiosin is a potent inhibitor of the Wnt/β-catenin pathway. Prodigiosin has antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer properties.
Specifikationer
Kemiskt namn eller material Prodigiosin
Fysisk form Solid
CAS 82-89-3
Färg Red
Kvantitet 100 μg
Anteckningar om renhetsgrad Research
Molekylformel C20H25N3O
För användning med (applikation) Cancer-programmed cell death
Synonym Prodigiosine
Löslighetsinformation DMSO : 25 mg/mL (77.30 mM; Need ultrasonic)
LEDER CCCCCC1=C/C(N=C1C)=C\C2=C(OC)C=C(C3=CC=CN3)N2
Molekylvikt (g/mol) 323.43
Formel vikt 323.43
Procent renhet 95.44%