Learn More
Profilin 1 Mouse anti-Human, Unconjugated, Clone: 3A12E8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67390-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16899985
  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
The Profilin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Profilin 1. This antibody recognizes human, mouse, rat antigen. The Profilin 1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
Profilin 1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence
3A12E8
Unconjugated
20 μL
5216
IgG2b