Läs mer
Profluthrin, TRC
Produktkod. 30422791
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422791 500 mg
30412834 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422791 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP755890500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 223419-20-3
Molekylformel C17 H18 F4 O2
Molekylvikt (g/mol) 330.32
Synonym Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(1-propenyl)-, (2,3,5,6-tetrafluoro-4-methylphenyl)methyl ester (9CI), 4-Methyl-2,3,5,6-tetrafluorobenzyl 2,2-dimethyl-3-(1-propenyl)cyclopropanecarboxylate, Fairytale, Profluthrin, [2,3,5,6-Tetrafluoro-4-methylphenyl]methyl 2,2-dimethyl-3-(1-propenyl)cyclopropanecarboxylate
IUPAC-namn (2,3,5,6-tetrafluoro-4-methylphenyl)methyl 2,2-dimethyl-3-prop-1-enylcyclopropane-1-carboxylate
LEDER CC=CC1C(C(=O)OCc2c(F)c(F)c(C)c(F)c2F)C1(C)C
Specifikationer
Kemiskt namn eller material Profluthrin
Kvantitet 500 mg
InChI formel InChI=1S/C17H18F4O2/c1-5-6-10-11(17(10,3)4)16(22)23-7-9-14(20)12(18)8(2)13(19)15(9)21/h5-6,10-11H,7H2,1-4H3
Formel vikt 330.1243