missing translation for 'onlineSavingsMsg'
Läs mer

Progesterone Acetate, TRC

Produktkod. 30422895
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422895 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30422895 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7559101G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 17-alpha-Hydroxyprogesterone 17-acetate
CAS 302-23-8
Molekylformel C23 H32 O4
Kvantitet 1 g
Synonym Hydroxyprogesterone Acetate, 3,20-Dioxopregn-4-en-17-yl Acetate, Megestrol Acetate Imp. K (Pharmeuropa), Medroxyprogesterone Acetate Imp. H (EP)
InChI formel InChI=1S/C23H32O4/c1-14(24)23(27-15(2)25)12-9-20-18-6-5-16-13-17(26)7-10-21(16,3)19(18)8-11-22(20,23)4/h13,18-20H,5-12H2,1-4H3/t18-,19+,20+,21+,22+,23+/m1/s1
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molekylvikt (g/mol) 372.5
Formel vikt 372.2301
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.