Läs mer
Proglumide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Proglumide |
| CAS | 6620-60-6 |
| Molekylformel | C18 H26 N2 O4 |
| Kvantitet | 1 g |
| Synonym | Pentanoic acid','4-(benzoylamino)-5-(dipropylamino)-5-oxo-, Glutaramic acid','4-benzamido-N,N-dipropyl-','DL- (8CI), Pentanoic acid','4-(benzoylamino)-5-(dipropylamino)-5-oxo-','(±)-, 4-(Benzoylamino)-5-(dipropylamino)-5-oxopentanoic acid, (±)-Proglumide, Binoside, CR 242, DL-4-Benzamido-N,N-dipropylglutaramic acid, DL-Proglumide, Gastrotopic, Midelid, Milid, Milide, Nulsa, Proglumide, Promid, Promide (parasympatholytic), Ulcutin, W 5219, Xyde, Xylamide, Xylamide (gastroprotective agent) |
| InChI formel | InChI=1S/C18H26N2O4/c1-3-12-20(13-4-2)18(24)15(10-11-16(21)22)19-17(23)14-8-6-5-7-9-14/h5-9,15H,3-4,10-13H2,1-2H3,(H,19,23)(H,21,22) |
| LEDER | CCCN(CCC)C(=O)C(CCC(=O)O)NC(=O)c1ccccc1 |
| IUPAC-namn | 4-benzamido-5-(dipropylamino)-5-oxopentanoic acid |
| Molekylvikt (g/mol) | 334.41 |
| Formel vikt | 334.1893 |