Läs mer
SI Analytics™ ProLab 2500 pH Set
Handla allt SI Analytics Produkter
Kalibreringspunkter (pH):
1 till 5
Beskrivning
- Including stand, power supply, A 162 IDS, buffer solutions and IDS analog adapter
- Analog pH/ORP sensors : Z600 IDS analog adapter (included in delivery)
- Three universal measuring channels
- Digital sensor recognition
- Antibacterial keyboard
- Measures pH, ORP, ISE, dissolved oxygen, and conductivity
- Any combination of the same or different measuring parameters
- Color backlit graphic display with CMC, QSC and channel display
- An adapter for conventional pH electrodes (DIN plug) included in the delivery
- The digital signal transfer eliminates interference, safely allocates calibration data, automatically transmits sensor data
- The Quality Sensor Control (QSC) icon provides precise information about the actual state of the electrode and therefore increases the operational safety
- Automatic, digital capture of all sensor data for retraceability of measured values
- Available user administration for the safe allocation of user and measuring results or sample and measuring results
- Data output to PC, USB memory stick or selected printers
Specifikationer
Specifikationer
| Kalibreringspunkter (konduktivitet) | 1 |
| Kalibreringspunkter (ISE) | 2 to 7 (requires adapter) |
| Kalibreringspunkter (pH) | 1 till 5 |
| Kanaler | 3 |
| Visningstyp | Color Graphic |
| Minne | Manual: 500 data sets; Automatic: 10000 data sets |
| Strömförsörjning | Universal |