Läs mer
Proligestone, TRC
Produktkod. 30415847
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30415847 5 mg
1 options
Produktkod. 30415847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7559605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Proligestone
CAS 23873-85-0
Molekylformel C24 H34 O4
Kvantitet 5 mg
Synonym Pregn-4-ene-3,20-dione','14,17-[propylidenebis(oxy)]-, Pregn-4-ene-3,20-dione','14,17-dihydroxy-','cyclic acetal with propionaldehyde (8CI), 1,4a-Ethano-4aH-phenanthro[1,2-d][1,3]dioxin','pregn-4-ene-3,20-dione deriv., 14,17-[Propylidenebis(oxy)]pregn-4-ene-3,20-dione, 14α,17α-Propylidenedioxypregn-4-ene-3,20-dione, Proligestone
InChI formel InChI=1S/C24H34O4/c1-5-20-27-23(15(2)25)12-13-24(28-20)19-7-6-16-14-17(26)8-10-21(16,3)18(19)9-11-22(23,24)4/h14,18-20H,5-13H2,1-4H3/t18-,19+,20?,21-,22+,23-,24+/m0/s1
LEDER CCC1O[C@@]23CC[C@](O1)(C(=O)C)[C@@]2(C)CC[C@H]4[C@H]3CCC5=CC(=O)CC[C@]45C
Molekylvikt (g/mol) 386.52
Formel vikt 386.2457