Läs mer
Promazine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Promazine hydrochloride |
| CAS | 53-60-1 |
| Molekylformel | C17 H20 N2 S . Cl H |
| Kvantitet | 2.5 g |
| Synonym | 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Promazine Hydrochloride, Chlorpromazine Hydrochloride Imp. C (EP) as Hydrochloride, 10H-Phenothiazine-10-propanamine','N,N-dimethyl-','hydrochloride (1:1), 10H-Phenothiazine-10-propanamine','N,N-dimethyl-','monohydrochloride (9CI), Phenothiazine','10-[3-(dimethylamino)propyl]-','monohydrochloride (8CI), 10-(γ-Dimethylamino-n-propyl)phenothiazine hydrochloride, Esparinal, Liranol, Prolactyl, Promazine monohydrochloride, Promwill, Propazin, Propazin (pharmaceutical), Propazine, Prozine, Sparine, Talofen |
| InChI formel | InChI=1S/C17H20N2S.ClH/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)20-17-11-6-4-9-15(17)19;/h3-6,8-11H,7,12-13H2,1-2H3;1H |
| LEDER | Cl.CN(C)CCCN1c2ccccc2Sc3ccccc13 |
| IUPAC-namn | N,N-dimethyl-3-phenothiazin-10-ylpropan-1-amine;hydrochloride |
| Molekylvikt (g/mol) | 320.88 |
| Formel vikt | 320.1114 |