Läs mer
Promethazine Sulfone, TRC
Produktkod. 30426710
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30426710 2.5 mg
1 options
Produktkod. 30426710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7570152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Promethazine Sulfone
CAS 13754-56-8
Molekylformel C17 H20 N2 O2 S
Kvantitet 2.5 mg
Synonym N,N,α-Trimethyl-10H-phenothiazine-10-ethanamine 5,5-Dioxide, Phenothiazine','10-[2-(dimethylamino)propyl]-','5,5-dioxide (8CI), 9,9-Dioxopromethazine, 9,9-Dioxypromethazin, Dioxopromethazine, Promethazine 5,5-dioxide, Prothanon, Promethazine Sulfone, N,N,α-Trimethyl-5,5-dioxide-10H-phenothiazine-10-ethanamine, 10-[2-(dimethylamino)propyl]-5,5-dioxidephenothiazine
InChI formel InChI=1S/C17H20N2O2S/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)22(20,21)17-11-7-5-9-15(17)19/h4-11,13H,12H2,1-3H3
LEDER CC(CN1c2ccccc2S(=O)(=O)c3ccccc13)N(C)C
IUPAC-namn 1-(5,5-dioxophenothiazin-10-yl)-N,N-dimethyl-propan-2-amine
Molekylvikt (g/mol) 316.42
Formel vikt 316.1245