Läs mer
Promethazine Sulfone, TRC
Produktkod. 30422529
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422529 25 mg
30421539 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP75701525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Promethazine Sulfone
CAS 13754-56-8
Molekylformel C17 H20 N2 O2 S
Kvantitet 25 mg
Synonym N,N,α-Trimethyl-10H-phenothiazine-10-ethanamine 5,5-Dioxide, Phenothiazine','10-[2-(dimethylamino)propyl]-','5,5-dioxide (8CI), 9,9-Dioxopromethazine, 9,9-Dioxypromethazin, Dioxopromethazine, Promethazine 5,5-dioxide, Prothanon, Promethazine Sulfone, N,N,α-Trimethyl-5,5-dioxide-10H-phenothiazine-10-ethanamine, 10-[2-(dimethylamino)propyl]-5,5-dioxidephenothiazine
InChI formel InChI=1S/C17H20N2O2S/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)22(20,21)17-11-7-5-9-15(17)19/h4-11,13H,12H2,1-3H3
LEDER CC(CN1c2ccccc2S(=O)(=O)c3ccccc13)N(C)C
IUPAC-namn 1-(5,5-dioxophenothiazin-10-yl)-N,N-dimethyl-propan-2-amine
Molekylvikt (g/mol) 316.42
Formel vikt 316.1245