Läs mer
Prometon, TRC
Produktkod. 30412831
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412831 25 mg
30410757 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412831 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP75703525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Prometon
CAS 1610-18-0
Molekylformel C10 H19 N5 O
Kvantitet 25 mg
Synonym Prometon, 1,3,5-Triazine-2,4-diamine','6-methoxy-N2,N4-bis(1-methylethyl)- (ACI), 1,3,5-Triazine-2,4-diamine','6-methoxy-N,N'-bis(1-methylethyl)- (9CI), 6-Methoxy-N2,N4-bis(1-methylethyl)-1,3,5-triazine-2,4-diamine (ACI), s-Triazine','2,4-bis(isopropylamino)-6-methoxy- (6CI','8CI), 2,4-Bis(isopropylamino)-6-methoxy-1,3,5-triazine, 2,4-Bis(isopropylamino)-6-methoxy-s-triazine, 2-Methoxy-4,6-bis(isopropylamino)-1,3,5-triazine, 2-Methoxy-4,6-bis(isopropylamino)-s-triazine, 4,6-Bis(isopropylamino)-2-methoxy-s-triazine, G 31435, Gesafram, Methoxypropazine, NSC 163048, Pramitol, Pramitol 5P, Prometon, Prometone
InChI formel InChI=1S/C10H19N5O/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7H,1-5H3,(H2,11,12,13,14,15)
LEDER COc1nc(NC(C)C)nc(NC(C)C)n1
IUPAC-namn 6-methoxy-2-N,4-N-di(propan-2-yl)-1,3,5-triazine-2,4-diamine
Molekylvikt (g/mol) 225.29
Formel vikt 225.159