Läs mer
Pronase E (Activity ≥ 7000 U/g), MedChemExpress
Beskrivning
Pronase E (Activity ≥ 7000 U/g) is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus and could digest protein into individual amino acids.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pronase E (Activity ≥ 7000 U/g) |
| Fysisk form | Solid |
| CAS | 9036-06-0 |
| Färg | Off-White |
| Kvantitet | 100 mg |
| Anteckningar om renhetsgrad | Research |
| Synonym | Pronase (Activity ≥ 7000 U/g) |
| Löslighetsinformation | DMSO : 50 mg/mL (Need ultrasonic) ∣H2O : 20 mg/mL (Need ultrasonic) |
| LEDER | [Pronase E (Activity ≥ 7000 U/g)] |
| Kvalitet | Research |