Läs mer
Propachlor, TRC
Produktkod. 30421725
Change view
Kvantitet:
100 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30421725 1 g -
30413071 100 mg -
30411732 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7573001G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1918-16-7
Molekylformel C11 H14 Cl N O
Molekylvikt (g/mol) 211.69
Synonym Acetanilide, 2-chloro-N-isopropyl- (8CI), 2-Chloro-N-(1-methylethyl)-N-phenylacetamide, 2-Chloro-N-isopropyl-N-phenylacetamide, 2-Chloro-N-isopropylacetanilide, Acilid, Albrass, Bexton, Bexton 4L, CP 31393, Kartex A, Muharicid, N-Isopropyl-2-chloroacetanilide, N-Isopropyl-α-chloroacetanilide, Niticid, Propachlor, Propachlore, Ramrod, Ramrod 65, Ramrod Flo, Satecid, α-Chloro-N-isopropylacetanilide
IUPAC-namn 2-chloro-N-phenyl-N-propan-2-ylacetamide
LEDER CC(C)N(C(=O)CCl)c1ccccc1
Specifikationer
Kemiskt namn eller material Propachlor
Kvantitet 1 g
InChI formel InChI=1S/C11H14ClNO/c1-9(2)13(11(14)8-12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3
Formel vikt 211.0764