Läs mer
Propafenone Dimer Impurity (Mixture of Diastereomers), TRC
Produktkod. 17974105
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974105 5 mg
17941805 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17974105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7575205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346603-80-2
Molekylformel C39H45NO6
Molekylvikt (g/mol) 623.78
Synonym 1,1'-[Propyliminobis[(2-hydroxypropane-3,1-diyl)oxy-2,1-phenylene]]bis(3-phenylpropan-1-one), Propafenone Imp. G (EP), 1-[2-[2-Hydroxy-3-[[2-hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propyl]-propylamino]propoxy]phenyl]-3-phenylpropan-1-one
IUPAC-namn 1-[2-[2-hydroxy-3-[[2-hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propyl]-propylamino]propoxy]phenyl]-3-phenylpropan-1-one
LEDER CCCN(CC(O)COc1ccccc1C(=O)CCc2ccccc2)CC(O)COc3ccccc3C(=O)CCc4ccccc4
Specifikationer
Kemiskt namn eller material Propafenone Dimer Impurity (Mixture of diastereomers)
Kvantitet 5 mg
InChI formel InChI=1S/C39H45NO6/c1-2-25-40(26-32(41)28-45-38-19-11-9-17-34(38)36(43)23-21-30-13-5-3-6-14-30)27-33(42)29-46-39-20-12-10-18-35(39)37(44)24-22-31-15-7-4-8-16-31/h3-20,32-33,41-42H,2,21-29H2,1H3
Formel vikt 623.3247