Läs mer
Propan-2-yl 2-Sulfanylacetate, TRC
Produktkod. 30010887
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010887 500 mg
30011465 100 mg
30005936 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010887 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB524470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7383-61-1
Molekylformel C5 H10 O2 S
Molekylvikt (g/mol) 134.19
Synonym 1-Methylethyl 2-Mercaptoacetate, Isopropyl 2-Mercaptoacetate, Isopropyl Mercaptoacetate, Isopropyl Thioglycolate, Thioglycolic Acid Isopropyl Ester;
IUPAC-namn isopropyl 2-mercaptoacetate
LEDER CC(C)OC(=O)CS
Specifikationer
Kemiskt namn eller material Propan-2-yl 2-Sulfanylacetate
Kvantitet 500 mg
InChI formel InChI=1S/C5H10O2S/c1-4(2)7-5(6)3-8/h4,8H,3H2,1-2H3
Formel vikt 134.0402