Läs mer
Propan-2-yl Ethane-1-sulfonate, TRC
Produktkod. 30012062
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012062 100 mg
30009484 50 mg
30008104 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012062 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB523895100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14245-62-6
Molekylformel C5H12O3S
Molekylvikt (g/mol) 152.21
Synonym 1-Methylethyl Ethanesulfonate, Isopropyl Ethanesulfonate
IUPAC-namn propan-2-yl ethanesulfonate
LEDER CCS(=O)(=O)OC(C)C
Specifikationer
Kemiskt namn eller material Propan-2-yl Ethane-1-sulfonate
Kvantitet 100 mg
InChI formel InChI=1S/C5H12O3S/c1-4-9(6,7)8-5(2)3/h5H,4H2,1-3H3
Formel vikt 152.0507