Läs mer
Propanil, TRC
Produktkod. 30135805
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135805 10 g
17988389 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135805 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76084010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 709-98-8
Molekylformel C9 H9 Cl2 N O
Molekylvikt (g/mol) 218.08
Synonym Propanamide, N-(3,4-dichlorophenyl)-, Propionanilide, 3',4'-dichloro- (6CI,8CI), N-(3,4-Dichlorophenyl)propanamide, 3',4'-Dichloropropionanilide, Chem Rice, DCPA, DPA, FW 734, N-(3,4-Dichlorophenyl)propionamide, NSC 31312, Propagrin 36, Propanex, Propanid, Propanide, Propanil, Propasint, Propionic acid 3,4-dichloroanilide, Riceshot, Riverside propanil 60DF, Rogue, Stam, Stam 480EC, Stam 80EDF, Stam F 34, Stam LV 10, Stam M 4, Stam Novel Flo 480, Stampede 360, Strel, Surcopur, Synpran N, Wham EZ
IUPAC-namn N-(3,4-dichlorophenyl)propanamide
LEDER CCC(=O)Nc1ccc(Cl)c(Cl)c1
Specifikationer
Kemiskt namn eller material Propanil
Kvantitet 10 g
InChI formel InChI=1S/C9H9Cl2NO/c1-2-9(13)12-6-3-4-7(10)8(11)5-6/h3-5H,2H2,1H3,(H,12,13)
Formel vikt 217.0061