Läs mer
Propantheline Bromide, LoGiCal
Produktkod. 30458258
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30458258 10 mg
1 options
Produktkod. 30458258 Leverantör LoGiCal Leverantörsnummer LGCFOR1450.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propantheline Bromide
CAS 50-34-0
Analyt- eller komponentnamn Propantheline Bromide
Frakt skick Ice pack (-20°C)
Molekylformel C23 H30 N O3 . Br
Certifikat och efterlevnad ISO 9001
Synonym Propantheline bromide, N-Methyl-N,N-bis(1-methylethyl)-2-[(9H-xanthen-9-ylcarbonyl)oxy]ethanaminium bromide, (2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate (6CI), 2-Propanaminium, N-methyl-N-(1-methylethyl)-N-[2-[(9H-xanthen-9-ylcarbonyl)oxy]ethyl]-, bromide (9CI), Ammonium, (2-hydroxyethyl)diisopropylmethyl-, bromide, xanthene-9-carboxylate (8CI), Corrigast, Ercorax, Ercotina, Ketaman, Kivatin, Neo-Metantyl, Neopepulsan, Pantas, Pantheline, Pervagal, Pro-Banthine, Pro-Gastron, Prodixamon, Propantel, SC 3171, Xanthene-9-carboxylic acid, ester with (2-hydroxyethyl)diisopropylmethylammonium bromide, beta-Diisopropylaminoethyl xanthene-9-carboxylate methobromide
InChI formel InChI=1S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22H,14-15H2,1-5H3;1H/q+1;/p-1
Lösningstyp Neat
LEDER [Br-].CC(C)[N+](C)(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C
IUPAC-namn methyl-di(propan-2-yl)-[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium;bromide
Industry Type Clinical Testing
Molekylvikt (g/mol) 448.39
Kvantitet 10 mg
Formel vikt 447.1409 g/mol