Läs mer
Propantheline bromide, MedChemExpress
Produktkod. 16468666
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16468666 100 mg
1 options
Produktkod. 16468666 Leverantör Medchem Express Leverantörsnummer HYB1188
MedChemExpress Propantheline bromide is an orally active mAChR antagonist. Propantheline bromide can be used in the research of smooth muscle dysfunction, excessive sweating, cramps or spasms of the stomach, intestines or bladder, and involuntary urination.
Propantheline bromide is an orally active mAChR antagonist. Propantheline bromide can be used in the research of smooth muscle dysfunction, excessive sweating, cramps or spasms of the stomach, intestines or bladder, and involuntary urination.
Specifikationer
Kemiskt namn eller material Propantheline bromide
CAS 50-34-0
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C23H30BrNO3
Kvantitet 100 mg
Löslighetsinformation DMSO : ≥ 100 mg/mL (223.02 mM) ∣H2O : 50 mg/mL (111.51 mM; Need ultrasonic)
LEDER CC([N+](C(C)C)(C)CCOC(C1C2=C(OC3=C1C=CC=C3)C=CC=C2)=O)C.[Br-]
Molekylvikt (g/mol) 448.39
Formel vikt 448.39
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation