Läs mer
Propantheline Bromide, TRC
Produktkod. 30425461
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30425461 25 mg
1 options
Produktkod. 30425461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76090025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propantheline Bromide
CAS 50-34-0
Molekylformel C23 H30 N O3 . Br
Kvantitet 25 mg
Synonym Propantheline bromide, N-Methyl-N,N-bis(1-methylethyl)-2-[(9H-xanthen-9-ylcarbonyl)oxy]ethanaminium bromide, (2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate (6CI), 2-Propanaminium','N-methyl-N-(1-methylethyl)-N-[2-[(9H-xanthen-9-ylcarbonyl)oxy]ethyl]-','bromide (9CI), Ammonium','(2-hydroxyethyl)diisopropylmethyl-','bromide','xanthene-9-carboxylate (8CI), Corrigast, Ercorax, Ercotina, Ketaman, Kivatin, Neo-Metantyl, Neopepulsan, Pantas, Pantheline, Pervagal, Pro-Banthine, Pro-Gastron, Prodixamon, Propantel, Propantheline bromide, SC 3171, Xanthene-9-carboxylic acid','ester with (2-hydroxyethyl)diisopropylmethylammonium bromide, β-Diisopropylaminoethyl xanthene-9-carboxylate methobromide
InChI formel InChI=1S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22H,14-15H2,1-5H3;1H/q+1;/p-1
LEDER [Br-].CC(C)[N+](C)(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C
IUPAC-namn methyl-di(propan-2-yl)-[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium;bromide
Molekylvikt (g/mol) 448.39
Formel vikt 447.1409