Läs mer
Propargite(Mixture of Diastereomers), TRC
Produktkod. 30137513
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137513 10 g
30134436 25 g
17918099 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137513 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76250010G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2312-35-8
Molekylformel C19 H26 O4 S
Molekylvikt (g/mol) 350.47
Synonym Sulfurous acid, 2-(p-tert-butylphenoxy)cyclohexyl 2-propynyl ester (7CI,8CI), Sulfurous acid, 2-[4-(1,1-dimethylethyl)phenoxy]cyclohexyl 2-propynyl ester (9CI), 2-(p-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite, 2-(p-tert-Butylphenoxy)cyclohexyl propargyl sulfite, Acarit 720EC, BPPS, Comite, Comite II, Cyclosulfyne, D 014, D 014 (ester), Kelaran, Naugatuck D 014, Omite, Omite 57E, Omite 85E, Propargil, Propargite, Sulfurous acid, 2-[4-(1,1-dimethylethyl)phenoxy]cyclohexyl 2-propyn-1-yl ester
IUPAC-namn [2-(4-tert-butylphenoxy)cyclohexyl] prop-2-ynyl sulfite
LEDER CC(C)(C)c1ccc(OC2CCCCC2OS(=O)OCC#C)cc1
Specifikationer
Kemiskt namn eller material Propargite
Kvantitet 10 g
InChI formel InChI=1S/C19H26O4S/c1-5-14-21-24(20)23-18-9-7-6-8-17(18)22-16-12-10-15(11-13-16)19(2,3)4/h1,10-13,17-18H,6-9,14H2,2-4H3
Formel vikt 350.1552