Learn More
Propargyl chloride, 98%, Thermo Scientific Chemicals
Catalog Number p-4813628
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
624-65-7
C3H3Cl
74.51
MFCD00000980
LJZPPWWHKPGCHS-UHFFFAOYSA-N
propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
12221
3-chloroprop-1-yne
ClCC#C
624-65-7
C3H3Cl
74.51
MFCD00000980
LJZPPWWHKPGCHS-UHFFFAOYSA-N
propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
12221
3-chloroprop-1-yne
ClCC#C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0300g/mL
Boiling Point 58.0°C to 61.0°C
Flash Point 19°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2Cl
Quantity 5 g
Merck Index 15,7924
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble.
Formula Weight 74.51
Percent Purity 98%
Chemical Name or Material Propargyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.