Learn More
IKA Propeller Stirrer, 3-Bladed
Overhead, propeller-shaped stirrer element with a flow-efficient design suitable for medium to high-speed mixing.
2853.22 SEK - 3410.99 SEK
Specifications
For Use With (Application) Mixing at medium to high speeds.
Type Propeller Stirrer, 3-bladed
View More Specs
Products 4
Product Code Brand Diameter (Metric) Fan Diameter (Metric) Shaft Length (Metric) Speed Price Quantity & Availability  
Product Code Brand Diameter (Metric) Fan Diameter (Metric) Shaft Length (Metric) Speed Price Quantity & Availability  
10622401 View Documents IKA
0001296000
45 mm 8 mm 350 mm 2000 rpm 2853.22 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
28-02-2020
Add to basket
10748341 View Documents IKA
0001295900
55 mm 8 mm 350 mm 2000 rpm 2971.21 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
28-02-2020
Add to basket
10602781 View Documents IKA
0000477700
140 mm 10 mm 550 mm 800 rpm Max. 2853.22 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
28-02-2020
Add to basket
10008220 View Documents IKA
0000477800
140 mm 10 mm 800 mm 400 rpm Max. 3410.99 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
28-02-2020
Add to basket
Description
Description
  • Propeller design stirrer
  • Three-bladed
  • Flow efficient design
  • For drawing material to be mixed from the bottom to the top
  • Minimum shearing forces
  • Used at medium to high speeds
Specifications
Specifications
Mixing at medium to high speeds.
Propeller Stirrer, 3-bladed