Läs mer
Propineb, MedChemExpress
Produktkod. 16475676
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16475676 100 mg
1 options
Produktkod. 16475676 Leverantör Medchem Express Leverantörsnummer HY119630
MedChemExpress Propineb (Zinc propylenebis) is a compound widely used in fruit and vegetables cultures, due to its large spectrum of activity against fungal plant diseases.
Propineb (Zinc propylenebis) is a compound widely used in fruit and vegetables cultures, due to its large spectrum of activity against fungal plant diseases.
Specifikationer
Kemiskt namn eller material Propineb
Fysisk form Solid
CAS 12071-83-9
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C5H8N2S4Zn2-
För användning med (applikation) Neuroscience-Neuromodulation
Synonym Zinc propylenebis(dithiocarbamate)
Löslighetsinformation DMSO : 33.33 mg/mL (115.02 mM; Need ultrasonic)
LEDER S=C([S-])NCC(C)NC1=[S][Zn+2][S-]1
Molekylvikt (g/mol) 289.77
Formel vikt 355.15