Läs mer
Propioamidine Hydrochloride, TRC
Produktkod. 30428784
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30428784 1 g
1 options
Produktkod. 30428784 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8390901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propioamidine Hydrochloride
CAS 3599-89-1
Molekylformel C3H8N2 • (HCl)
Kvantitet 1 g
Synonym Propionamidine','hydrochloride, Propanamidine hydrochloride, Propanimidamide hydrochloride, Propioamidine hydrochloride, Propionimidamide hydrochloride
InChI formel InChI=1S/C3H8N2.ClH/c1-2-3(4)5;/h2H2,1H3,(H3,4,5);1H
LEDER N=C(CC)N.Cl
IUPAC-namn propionimidamide hydrochloride
Molekylvikt (g/mol) 108.57
Formel vikt 108.05