Läs mer
Propiolic Acid Sodium Salt (90%), TRC
Produktkod. 30414280
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30414280 500 mg
1 options
Produktkod. 30414280 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP786330500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propiolic Acid Sodium Salt
CAS 920-38-7
Molekylformel C3HNaO2
Kvantitet 500 mg
Synonym Acetylenecarboxylic Acid Sodium Salt, 2-Propynoic Acid Sodium Salt, Sodium Propiolate, Sodium Propynoate
InChI formel InChI=1S/C3H2O2.Na/c1-2-3(4)5;/h1H,(H,4,5);/q;+1/p-1
LEDER [Na+].[O-]C(=O)C#C
IUPAC-namn sodium;prop-2-ynoate
Molekylvikt (g/mol) 92.03
Formel vikt 91.9874