Learn More
Propionamidoxime, 97%, Thermo Scientific Chemicals
713.21 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
Propionamidoxime is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propionamidoxime is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Propionamidoxime is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications
Quantity | 250 mg |
Sensitivity | Hygroscopic |
Solubility Information | Slightly soluble in water. |
Formula Weight | 88.11 |
Percent Purity | 97% |
Chemical Name or Material | Propionamidoxime |