Läs mer
Propionyl Coenzyme A Lithium Salt (>85%), TRC
Produktkod. 30427025
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30427025 5 mg
1 options
Produktkod. 30427025 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8390435MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propionyl Coenzyme A Lithium Salt
CAS 108321-21-7
Molekylformel C24 H40 N7 O17 P3 S
Kvantitet 5 mg
Synonym Coenzyme A S-propanoate Lithium Salt, S-(2-(3-((2R)-4-(((((((2S,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-hydroxy-3-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)oxy)-2-hydroxy-3,3-dimethylbutanamido)propanamido)ethyl) Propanethioate
InChI formel InChI=1S/C24H40N7O17P3S/c1-4-15(33)52-8-7-26-14(32)5-6-27-22(36)19(35)24(2,3)10-45-51(42,43)48-50(40,41)44-9-13-18(47-49(37,38)39)17(34)23(46-13)31-12-30-16-20(25)28-11-29-21(16)31/h11-13,17-19,23,34-35H,4-10H2,1-3H3,(H,26,32)(H,27,36)(H,40,41)(H,42,43)(H
LEDER CCC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23
IUPAC-namn S-[2-[3-[[(2R)-4-[[[(2S,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] propanethioate
Molekylvikt (g/mol) 823.597
Formel vikt 823.141