Läs mer
Propionyl-L-carnitine, TRC
Produktkod. 30426013
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30426013 5 mg -
30413888 10 mg -
30426567 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426013 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2271905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20064-19-1
Molekylformel C10H19NO4
Molekylvikt (g/mol) 217.26
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-, inner salt, (2R)- (9CI, ACI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-, inner salt, (R)- (ZCI), Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, hydroxide, inner salt, propionate, (-)- (8CI), (-)-Propionylcarnitine, L-Carnitine propionyl ester, L-Propionylcarnitine, C3-Carnitine, Levocarnitine propil, O-Propionyl-L-carnitine, Propanoyl-L-carnitine, Propionyl-L-carnitine, Propionylcarnitine, (2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium inner salt
IUPAC-namn (3R)-3-propanoyloxy-4-(trimethylazaniumyl)butanoate
LEDER CCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C
Specifikationer
Kemiskt namn eller material Propionyl-L-carnitine
Kvantitet 5 mg
InChI formel InChI=1S/C10H19NO4/c1-5-10(14)15-8(6-9(12)13)7-11(2,3)4/h8H,5-7H2,1-4H3/t8-/m1/s1
Formel vikt 217.1314