Läs mer
Propiverine Hydrochloride, TRC
Produktkod. 30414429
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414429 1 g
30422318 500 mg
30417751 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30414429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8280001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54556-98-8
Molekylformel C23 H29 N O3 . Cl H
Molekylvikt (g/mol) 403.94
Synonym α-phenyl-α-propoxy-Benzeneacetic acid 1-methyl-4-piperidinyl ester hydrochloride (1:1), Acetic acid, diphenylpropoxy-, 1-methyl-4-piperidyl ester, hydrochloride (7CI), Benzeneacetic acid, α-phenyl-α-propoxy-, 1-methyl-4-piperidinyl ester, hydrochloride (9CI), Mictonetten, Mictonorm, P 4, P 4 (anticholinergic), Pollarine, Propiverine hydrochloride
IUPAC-namn (1-methylpiperidin-4-yl) 2,2-diphenyl-2-propoxyacetate;hydrochloride
LEDER Cl.CCCOC(C(=O)OC1CCN(C)CC1)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Propiverine, Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C23H29NO3.ClH/c1-3-18-26-23(19-10-6-4-7-11-19,20-12-8-5-9-13-20)22(25)27-21-14-16-24(2)17-15-21;/h4-13,21H,3,14-18H2,1-2H3;1H
Formel vikt 403.1914
Procent renhet >95%