Läs mer
Propoxyphenyl Thioaildenafil, TRC
Produktkod. 30412642
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412642 50 mg
30421119 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP83164050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856190-49-3
Molekylformel C24H34N6O3S2
Molekylvikt (g/mol) 518.7
Synonym rel-5-[5-[[(3R,5S)-3,5-Dimethyl-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, (3R,5S)-rel-1-[[3-(4,7-Dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxyphenyl]sulfonyl]-3,5-dimethyl-piperazine, 5-(5-(((3R,5S)-3,5-Dimethylpiperazin-1-yl)sulfonyl)-2-propoxyphenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidine-7(4H)-thione
LEDER S=C1C2=C(C(CCC)=NN2C)NC(C3=CC(S(N4C[C@@H](C)N[C@@H](C)C4)(=O)=O)=CC=C3OCCC)=N1
Specifikationer
Kemiskt namn eller material Propoxyphenyl Thioaildenafil
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H34N6O3S2/c1-6-8-19-21-22(29(5)28-19)24(34)27-23(26-21)18-12-17(9-10-20(18)33-11-7-2)35(31,32)30-13-15(3)25-16(4)14-30/h9-10,12,15-16,25H,6-8,11,13-14H2,1-5H3,(H,26,27,34)/t15-,16+
Formel vikt 518.2134
Procent renhet >95%