Läs mer
Propoxyphenyl-thiohydroxyhomosildenafil, TRC
Produktkod. 30005356
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005356 5 mg
30011264 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8316305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 479073-90-0
Molekylformel C24H34N6O4S2
Molekylvikt (g/mol) 534.69
Synonym 1-Piperazineethanol, 4-[[3-(4,7-dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxyphenyl]sulfonyl]- (9CI), 1,6-Dihydro-5-[5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, Propoxyphenyl-thiohydroxyhomosildenafil
IUPAC-namn 5-[5-[4-(2-hydroxyethyl)piperazin-1-yl]sulfonyl-2-propoxyphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidine-7-thione
LEDER CCCOc1ccc(cc1C2=NC(=S)c3c(N2)c(CCC)nn3C)S(=O)(=O)N4CCN(CCO)CC4
Specifikationer
Kemiskt namn eller material Propoxyphenyl-Thiohydroxyhomosildenafil
Kvantitet 5 mg
InChI formel InChI=1S/C24H34N6O4S2/c1-4-6-19-21-22(28(3)27-19)24(35)26-23(25-21)18-16-17(7-8-20(18)34-15-5-2)36(32,33)30-11-9-29(10-12-30)13-14-31/h7-8,16,31H,4-6,9-15H2,1-3H3,(H,25,26,35)
Formel vikt 534.2083