Läs mer
Propyl 2-Hydroxyacetic Acid Ester, TRC
Produktkod. 30427679
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30427679 250 mg
1 options
Produktkod. 30427679 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP835825250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propyl 2-Hydroxyacetic Acid Ester
CAS 90357-58-7
Molekylformel C5H10O3
Kvantitet 250 mg
Synonym Propyl Hydroxyacetic Acid Ester, Propyl Glycolate
InChI formel InChI=1S/C5H10O3/c1-2-3-8-5(7)4-6/h6H,2-4H2,1H3
LEDER CCCOC(=O)CO
IUPAC-namn propyl 2-hydroxyacetate
Molekylvikt (g/mol) 118.13
Formel vikt 118.063