Läs mer
Propyl Acetoacetate, TRC
Produktkod. 30427678
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30427678 1 g
1 options
Produktkod. 30427678 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8336051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propyl Acetoacetate
CAS 1779-60-8
Molekylformel C13 H16 N4 O6 . H Cl
Kvantitet 1 g
Synonym 3-Oxo-butanoic Acid Propyl Ester, Acetoacetic Acid Propyl Ester, NSC 42862, Propyl 3-Oxobutanoate, n-Propyl Acetoacetate
InChI formel InChI=1S/C13H16N4O6.ClH/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15;/h1-2,7,11H,3-6,8-9H2;1H/b14-7+;
LEDER Cl.[O-][N+](=O)c1oc(\C=N\N2CC(CN3CCOCC3)OC2=O)cc1
IUPAC-namn 5-(morpholin-4-ylmethyl)-3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one;hydrochloride
Molekylvikt (g/mol) 360.75
Formel vikt 360.084