Läs mer
Propyl Benzoate, TRC
Produktkod. 30426754
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30426754 5 g
1 options
Produktkod. 30426754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1092955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propyl Benzoate
CAS 2315-68-6
Molekylformel C10 H12 O2
Kvantitet 5 g
Synonym Propyl Benzoate, Benzoic acid','propyl ester, NSC 229333, Propyl benzoate, n-Propyl benzoate
InChI formel InChI=1S/C10H12O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3
LEDER CCCOC(=O)c1ccccc1
IUPAC-namn propyl benzoate
Molekylvikt (g/mol) 164.201
Formel vikt 164.0837